Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGGTCAGGCACAACTACCTCAAA[C/T]GCAAACTCCGTGTCCCACCATGGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602142 | ||||||||||||||||||||
Literature Links: |
PLCD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PLCD1 - phospholipase C delta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130964.1 | 2454 | Silent Mutation | GCA,GCG | A,A 710 | NP_001124436.1 | |
NM_006225.3 | 2454 | Silent Mutation | GCA,GCG | A,A 689 | NP_006216.2 | |
XM_017006622.1 | 2454 | Silent Mutation | GCA,GCG | A,A 631 | XP_016862111.1 |
VILL - villin like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015873.3 | 2454 | Intron | NP_056957.3 | |||
XM_005265191.3 | 2454 | Intron | XP_005265248.1 | |||
XM_005265192.4 | 2454 | Intron | XP_005265249.1 | |||
XM_006713184.3 | 2454 | Intron | XP_006713247.1 | |||
XM_011533770.2 | 2454 | Intron | XP_011532072.1 | |||
XM_011533771.1 | 2454 | Intron | XP_011532073.1 | |||
XM_011533773.2 | 2454 | Intron | XP_011532075.1 |