Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCAGAGTGGGAGGTTTGGTTTAGC[A/G]GGGGCCAACTAGGGTCATTTGCCAA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 609556 | |||||||||||||||||||||||
Literature Links: |
ATP13A4 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ATP13A4 - ATPase 13A4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032279.3 | 3456 | Missense Mutation | CCG,CTG | P,L 1166 | NP_115655.2 | |
XM_011513232.2 | 3456 | Intron | XP_011511534.1 | |||
XM_017007318.1 | 3456 | Missense Mutation | CCG,CTG | P,L 1057 | XP_016862807.1 | |
XM_017007319.1 | 3456 | Intron | XP_016862808.1 |