Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGCCCCCTCACACATCAACCCCA[C/T]ACGCCTCCGCTGCACCCTGCCCTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 238310 MIM: 611516 MIM: 165390 MIM: 600690 | ||||||||||||||||||||
Literature Links: |
AMT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AMT - aminomethyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000481.3 | 1057 | Missense Mutation | ATG,GTG | M,V 321 | NP_000472.2 | |
NM_001164710.1 | 1057 | Missense Mutation | ATG,GTG | M,V 277 | NP_001158182.1 | |
NM_001164711.1 | 1057 | Missense Mutation | ATG,GTG | M,V 265 | NP_001158183.1 | |
NM_001164712.1 | 1057 | Missense Mutation | ATG,GTG | M,V 321 | NP_001158184.1 |
NICN1 - nicolin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RHOA - ras homolog family member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TCTA - T-cell leukemia translocation altered | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |