Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCAGGTGCCGCTTCTGCTCGGT[A/G]GCTGCGTCAATGTCCCCCAGCCGCA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 606738 | |||||||||||||||||||||||
Literature Links: |
OSBPL10 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
OSBPL10 - oxysterol binding protein like 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001174060.1 | 1994 | Silent Mutation | GCC,GCT | A,A 653 | NP_001167531.1 | |
NM_017784.4 | 1994 | Silent Mutation | GCC,GCT | A,A 717 | NP_060254.2 | |
XM_005264843.3 | 1994 | Silent Mutation | GCC,GCT | A,A 717 | XP_005264900.1 | |
XM_005264844.4 | 1994 | Silent Mutation | GCC,GCT | A,A 653 | XP_005264901.1 | |
XM_005264845.1 | 1994 | Silent Mutation | GCC,GCT | A,A 381 | XP_005264902.1 | |
XM_011533326.2 | 1994 | Silent Mutation | GCC,GCT | A,A 580 | XP_011531628.1 | |
XM_017005667.1 | 1994 | Silent Mutation | GCC,GCT | A,A 580 | XP_016861156.1 | |
XM_017005668.1 | 1994 | Silent Mutation | GCC,GCT | A,A 580 | XP_016861157.1 | |
XM_017005669.1 | 1994 | Silent Mutation | GCC,GCT | A,A 544 | XP_016861158.1 | |
XM_017005670.1 | 1994 | Silent Mutation | GCC,GCT | A,A 499 | XP_016861159.1 |