Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGAAAATATTTACTATTTTTAGA[A/T]AGAAGTTTTCCCATTTCCAGAAGTT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 611103 | |||||||||||||||||||||||
Literature Links: |
ACAD9 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ACAD9 - acyl-CoA dehydrogenase family member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014049.4 | 354 | Missense Mutation | AAA,ATA | K,I 51 | NP_054768.2 | |
XM_017006264.1 | 354 | UTR 5 | XP_016861753.1 | |||
XM_017006265.1 | 354 | Intron | XP_016861754.1 |