Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTCCTCTTCCTTGGGTTCTGGAC[A/G]CCTTACAACATAGTGCTCTTCCTAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604836 | ||||||||||||||||||||
Literature Links: |
CCR4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCR4 - C-C motif chemokine receptor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005508.4 | 983 | Silent Mutation | ACA,ACG | T,T 256 | NP_005499.1 | |
XM_017005687.1 | 983 | Silent Mutation | ACA,ACG | T,T 256 | XP_016861176.1 |
LOC105377022 - uncharacterized LOC105377022 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |