Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTTGGTCAGAAAGTGTAGATACC[A/G]ATGGATACTCTTCAGTAGCTGCAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZBTB11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZBTB11 - zinc finger and BTB domain containing 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014415.3 | 4635 | Missense Mutation | TCG,TTG | S,L 1007 | NP_055230.2 | |
XM_011512689.2 | 4635 | Missense Mutation | TCG,TTG | S,L 942 | XP_011510991.1 | |
XM_011512690.2 | 4635 | Missense Mutation | TCG,TTG | S,L 723 | XP_011510992.1 |