Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACTGTTCCAATTAAAGATAATTT[C/T]GAATTAGAGGTACTTCAGGCACAAT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 606605 MIM: 615808 | |||||||||||||||||||||||
Literature Links: |
ATRIP PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ATRIP - ATR interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271022.1 | 519 | UTR 5 | NP_001257951.1 | |||
NM_001271023.1 | 519 | Silent Mutation | TTC,TTT | F,F 19 | NP_001257952.1 | |
NM_032166.3 | 519 | Silent Mutation | TTC,TTT | F,F 112 | NP_115542.2 | |
NM_130384.2 | 519 | Silent Mutation | TTC,TTT | F,F 112 | NP_569055.1 |
CCDC51 - coiled-coil domain containing 51 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMA7 - translation machinery associated 7 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |