Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAAAAAAGGGTGTTAGTATTTTT[A/G]TCTTGGACACTCTCTTCCAAATCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602075 | ||||||||||||||||||||
Literature Links: |
SATB1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SATB1 - SATB homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001131010.3 | 2483 | Silent Mutation | GAC,GAT | D,D 734 | NP_001124482.1 | |
NM_001195470.2 | 2483 | Silent Mutation | GAC,GAT | D,D 766 | NP_001182399.1 | |
NM_001322871.1 | 2483 | Silent Mutation | GAC,GAT | D,D 766 | NP_001309800.1 | |
NM_001322872.1 | 2483 | Silent Mutation | GAC,GAT | D,D 734 | NP_001309801.1 | |
NM_001322873.1 | 2483 | Silent Mutation | GAC,GAT | D,D 734 | NP_001309802.1 | |
NM_001322874.1 | 2483 | Silent Mutation | GAC,GAT | D,D 734 | NP_001309803.1 | |
NM_001322875.1 | 2483 | Silent Mutation | GAC,GAT | D,D 734 | NP_001309804.1 | |
NM_001322876.1 | 2483 | Silent Mutation | GAC,GAT | D,D 662 | NP_001309805.1 | |
NM_002971.5 | 2483 | Silent Mutation | GAC,GAT | D,D 734 | NP_002962.1 | |
XM_011533988.2 | 2483 | Silent Mutation | GAC,GAT | D,D 766 | XP_011532290.1 | |
XM_011533989.2 | 2483 | Silent Mutation | GAC,GAT | D,D 766 | XP_011532291.1 | |
XM_011533990.2 | 2483 | Intron | XP_011532292.1 |