Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTGCCCGACGCACCAGGGTGCCC[C/T]GCCACCAGGCCTGGATCTTGGTGGC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
IQCF1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
IQCF1 - IQ motif containing F1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152397.2 | 337 | Missense Mutation | CAG,CGG | Q,R 80 | NP_689610.2 | |
XM_011533366.2 | 337 | Missense Mutation | CAG,CGG | Q,R 45 | XP_011531668.1 | |
XM_017005729.1 | 337 | Missense Mutation | CAG,CGG | Q,R 45 | XP_016861218.1 |