Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTCTGTCGCAAGACCTTGGAGAA[A/G]GCCAGCTTCATGTGCTGGTCCACTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 146691 MIM: 612911 | ||||||||||||||||||||
Literature Links: |
IMPDH2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IMPDH2 - inosine monophosphate dehydrogenase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NDUFAF3 - NADH:ubiquinone oxidoreductase complex assembly factor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
QRICH1 - glutamine rich 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320580.1 | 2356 | Silent Mutation | GCC,GCT | A,A 646 | NP_001307509.1 | |
NM_001320581.1 | 2356 | Silent Mutation | GCC,GCT | A,A 646 | NP_001307510.1 | |
NM_001320582.1 | 2356 | Silent Mutation | GCC,GCT | A,A 646 | NP_001307511.1 | |
NM_001320583.1 | 2356 | Silent Mutation | GCC,GCT | A,A 646 | NP_001307512.1 | |
NM_001320584.1 | 2356 | Silent Mutation | GCC,GCT | A,A 646 | NP_001307513.1 | |
NM_001320585.1 | 2356 | Silent Mutation | GCC,GCT | A,A 646 | NP_001307514.1 | |
NM_017730.3 | 2356 | Silent Mutation | GCC,GCT | A,A 646 | NP_060200.2 | |
NM_198880.2 | 2356 | Silent Mutation | GCC,GCT | A,A 646 | NP_942581.1 | |
XM_011533863.1 | 2356 | Silent Mutation | GCC,GCT | A,A 646 | XP_011532165.1 |