Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTTCACATTGGGCATTATCAATA[A/G]GATTGTCCATAAGGCTTACAAATCT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 607180 | |||||||||||||||||||||||
Literature Links: |
LRRC2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
LRRC2 - leucine rich repeat containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024512.4 | 1346 | Missense Mutation | CCT,CTT | P,L 318 | NP_078788.2 | |
XM_011534110.2 | 1346 | Missense Mutation | CCT,CTT | P,L 318 | XP_011532412.1 | |
XM_017007177.1 | 1346 | Intron | XP_016862666.1 |