Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGGACACGCTCAAGACCAGCAAC[C/T]CTCTGTGAGTGGGGGAAGCATGGCT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 601024 | |||||||||||||||||||||||
Literature Links: |
ABCF3 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ABCF3 - ATP binding cassette subfamily F member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018358.2 | 579 | Missense Mutation | CCT,TCT | P,S 148 | NP_060828.2 | |
XM_011512959.2 | 579 | UTR 5 | XP_011511261.1 |
AP2M1 - adaptor related protein complex 2 mu 1 subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |