Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCCTTACCGAGGTCTTGGCGACG[C/T]TGCCGCGCTGGGTGATGTTCTTGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609234 | ||||||||||||||||||||
Literature Links: |
EIF2A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EIF2A - eukaryotic translation initiation factor 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319043.1 | 1239 | Intron | NP_001305972.1 | |||
NM_001319044.1 | 1239 | Intron | NP_001305973.1 | |||
NM_001319045.1 | 1239 | Intron | NP_001305974.1 | |||
NM_001319046.1 | 1239 | Intron | NP_001305975.1 | |||
NM_032025.4 | 1239 | Intron | NP_114414.2 | |||
XM_011513224.1 | 1239 | Intron | XP_011511526.1 |
SERP1 - stress-associated endoplasmic reticulum protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014445.3 | 1239 | Missense Mutation | AAC,AGC | N,S 23 | NP_055260.1 | |
XM_011512698.2 | 1239 | Missense Mutation | AAC,AGC | N,S 23 | XP_011511000.1 |