Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTAGTTTCCAGCTAACAAGCCTCA[C/T]TCAGCAAGTGGTTTATAGTAACAAG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
C3orf33 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
C3orf33 - chromosome 3 open reading frame 33 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308229.1 | Intron | NP_001295158.1 | ||||
NM_173657.2 | Intron | NP_775928.1 | ||||
XM_005247379.4 | Intron | XP_005247436.1 | ||||
XM_011512710.2 | Intron | XP_011511012.1 | ||||
XM_011512711.2 | Intron | XP_011511013.1 |