Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTATTCCGAAACAAGTCTCGATTTC[A/G]TTCTGCAAACAATGTCAAAGGTACA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 611021 | |||||||||||||||||||||||
Literature Links: |
NMD3 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
NMD3 - NMD3 ribosome export adaptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320227.1 | 299 | Silent Mutation | TCA,TCG | S,S 53 | NP_001307156.1 | |
NM_015938.4 | 299 | Intron | NP_057022.2 | |||
XM_005247511.1 | 299 | Silent Mutation | TCA,TCG | S,S 53 | XP_005247568.1 | |
XM_005247512.1 | 299 | Silent Mutation | TCA,TCG | S,S 53 | XP_005247569.1 |