Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGACACCGATTGTGCCTAGAGTAA[A/C]CTGAAAAAATACGATTATTCTTAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613974 | ||||||||||||||||||||
Literature Links: |
DDX60 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DDX60 - DEXD/H-box helicase 60 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017631.5 | 5117 | Missense Mutation | GTT,TTT | V,F 1609 | NP_060101.3 | |
XM_011532103.2 | 5117 | Missense Mutation | GTT,TTT | V,F 1609 | XP_011530405.1 | |
XM_011532104.2 | 5117 | Intron | XP_011530406.1 | |||
XM_017008383.1 | 5117 | Intron | XP_016863872.1 | |||
XM_017008384.1 | 5117 | Intron | XP_016863873.1 |