Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTCTGTTTCCTGGATAAGGTGGT[C/T]GTGAAAACTAAGACCGAGTATGAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607645 | ||||||||||||||||||||
Literature Links: |
NSG1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NSG1 - neuron specific gene family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040101.1 | 372 | Silent Mutation | GTC,GTT | V,V 45 | NP_001035190.1 | |
NM_001287763.1 | 372 | Silent Mutation | GTC,GTT | V,V 45 | NP_001274692.1 | |
NM_001287764.1 | 372 | Silent Mutation | GTC,GTT | V,V 18 | NP_001274693.1 | |
NM_014392.4 | 372 | Silent Mutation | GTC,GTT | V,V 45 | NP_055207.1 | |
XM_017008022.1 | 372 | Silent Mutation | GTC,GTT | V,V 45 | XP_016863511.1 |