Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTAAGAGATGACCTGATCCAGGAT[A/G]TTGAAATACAGATTATTTCAAGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615912 MIM: 611355 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GSTCD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GSTCD - glutathione S-transferase C-terminal domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031720.3 | 289 | Missense Mutation | ATT,GTT | I,V 71 | NP_001026890.2 | |
NM_024751.3 | 289 | Intron | NP_079027.2 | |||
XM_005263222.3 | 289 | Missense Mutation | ATT,GTT | I,V 71 | XP_005263279.1 | |
XM_011532248.2 | 289 | Missense Mutation | ATT,GTT | I,V 71 | XP_011530550.1 | |
XM_011532249.2 | 289 | Missense Mutation | ATT,GTT | I,V 71 | XP_011530551.1 | |
XM_011532252.2 | 289 | Missense Mutation | ATT,GTT | I,V 71 | XP_011530554.1 |
INTS12 - integrator complex subunit 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |