Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGTGGAAATCAGCCTGTGATGTG[A/T]GGTGGTGGGCCAGCACCTAGAAGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114105 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP3CA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPP3CA - protein phosphatase 3 catalytic subunit alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000944.4 | 3707 | UTR 3 | NP_000935.1 | |||
NM_001130691.1 | 3707 | UTR 3 | NP_001124163.1 | |||
NM_001130692.1 | 3707 | UTR 3 | NP_001124164.1 | |||
XM_017008365.1 | 3707 | UTR 3 | XP_016863854.1 |