Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACGTGCACTCGGTAGGTGGTTAC[C/G]TTCATCACCCACGTGTCTGTCACAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602422 MIM: 605303 MIM: 615975 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLBP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLBP - stem-loop binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TACC3 - transforming acidic coiled-coil containing protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM129 - transmembrane protein 129 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127266.1 | 1039 | Missense Mutation | AAC,AAG | N,K 173 | NP_001120738.1 | |
NM_138385.3 | 1039 | Missense Mutation | AAC,AAG | N,K 173 | NP_612394.1 | |
XM_005248038.4 | 1039 | Missense Mutation | AAC,AAG | N,K 100 | XP_005248095.1 | |
XM_005248039.4 | 1039 | Missense Mutation | AAC,AAG | N,K 100 | XP_005248096.1 | |
XM_011513594.2 | 1039 | Missense Mutation | AAC,AAG | N,K 100 | XP_011511896.1 | |
XM_017008834.1 | 1039 | Missense Mutation | AAC,AAG | N,K 100 | XP_016864323.1 |