Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGTGCTTACTCACTGAGGGTACC[A/G]TTGCAGCAATTGGTATCATCAACAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610428 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COX18 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COX18 - COX18, cytochrome c oxidase assembly factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297732.1 | 931 | Missense Mutation | ACG,ATG | T,M 274 | NP_001284661.1 | |
NM_001297733.1 | 931 | Missense Mutation | ACG,ATG | T,M 122 | NP_001284662.1 | |
NM_001300729.1 | 931 | Missense Mutation | ACG,ATG | T,M 276 | NP_001287658.1 | |
NM_173827.3 | 931 | Missense Mutation | ACG,ATG | T,M 273 | NP_776188.1 | |
XM_005265680.4 | 931 | Missense Mutation | ACG,ATG | T,M 274 | XP_005265737.1 | |
XM_011531878.2 | 931 | Missense Mutation | ACG,ATG | T,M 123 | XP_011530180.1 | |
XM_017008045.1 | 931 | Missense Mutation | ACG,ATG | T,M 273 | XP_016863534.1 |