Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATCTTCGACAAGCACAGGTGACT[C/T]TCATAGACGCTACAACTTGCAATGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610399 | ||||||||||||||||||||
Literature Links: |
TMPRSS11E PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMPRSS11E - transmembrane protease, serine 11E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014058.3 | 1070 | Missense Mutation | CTC,TTC | L,F 336 | NP_054777.2 | |
XM_011531896.2 | 1070 | Missense Mutation | CTC,TTC | L,F 258 | XP_011530198.1 |