Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGGCTGTGGCACATGAAAAGGAC[A/G]TCCGTGTGTGTCTAGTGGAGAAAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 231675 | ||||||||||||||||||||
Literature Links: |
ETFDH PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ETFDH - electron transfer flavoprotein dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281737.1 | 507 | Missense Mutation | ATC,GTC | I,V 51 | NP_001268666.1 | |
NM_001281738.1 | 507 | Missense Mutation | ATC,GTC | I,V 37 | NP_001268667.1 | |
NM_004453.3 | 507 | Missense Mutation | ATC,GTC | I,V 98 | NP_004444.2 |