Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCACCGACCCCGTGAGCAAGACTT[C/T]CTCTCCGGAAGCTCTGGCCGTGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TRIML2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRIML2 - tripartite motif family like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303419.1 | 1306 | Missense Mutation | AAA,GAA | K,E 363 | NP_001290348.1 | |
NM_173553.2 | 1306 | UTR 3 | NP_775824.2 | |||
XM_011531737.2 | 1306 | Missense Mutation | AAA,GAA | K,E 338 | XP_011530039.1 | |
XM_011531738.1 | 1306 | Missense Mutation | AAA,GAA | K,E 338 | XP_011530040.1 | |
XM_011531739.1 | 1306 | Missense Mutation | AAA,GAA | K,E 246 | XP_011530041.1 | |
XM_017007882.1 | 1306 | Missense Mutation | AAA,GAA | K,E 338 | XP_016863371.1 |