Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTTCCTTCTCTTTGAACCTTGCA[C/T]GCTTGCCATTAACACCTTGGTGCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604724 | ||||||||||||||||||||
Literature Links: |
HPSE PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HPSE - heparanase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098540.2 | 1182 | Missense Mutation | ATG,GTG | M,V 423 | NP_001092010.1 | |
NM_001166498.2 | 1182 | Missense Mutation | ATG,GTG | M,V 349 | NP_001159970.1 | |
NM_001199830.1 | 1182 | Missense Mutation | ATG,GTG | M,V 365 | NP_001186759.1 | |
NM_006665.5 | 1182 | Missense Mutation | ATG,GTG | M,V 423 | NP_006656.2 |