Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTTTATTTTTCAGGGCTTCCTAT[A/G]ATACCTCTGCTCCAAATGCAAAACG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616504 | ||||||||||||||||||||
Literature Links: |
LOC101929741 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC101929741 - uncharacterized LOC101929741 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
METTL14 - methyltransferase like 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020961.3 | 326 | Missense Mutation | AAT,GAT | N,D 56 | NP_066012.1 | |
XM_005263166.4 | 326 | Missense Mutation | AAT,GAT | N,D 56 | XP_005263223.1 | |
XM_011532162.2 | 326 | Missense Mutation | AAT,GAT | N,D 56 | XP_011530464.1 |