Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCACCATAAGCTCTAATGCGTCGG[C/T]GGCCAAATACTTGGCAGTTTTGCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610669 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM193A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM193A - family with sequence similarity 193 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNIP2 - TNFAIP3 interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161527.1 | 1177 | Missense Mutation | ACC,GCC | T,A 257 | NP_001154999.1 | |
NM_001292016.1 | 1177 | Missense Mutation | ACC,GCC | T,A 281 | NP_001278945.1 | |
NM_024309.3 | 1177 | Missense Mutation | ACC,GCC | T,A 364 | NP_077285.3 |