Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTAAAACTCTAGGTCAGAGTCTCA[A/G]GATCGTTGGTGGGACAGAAGTAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610399 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMPRSS11E PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMPRSS11E - transmembrane protease, serine 11E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014058.3 | 636 | Missense Mutation | AAG,AGG | K,R 191 | NP_054777.2 | |
XM_011531896.2 | 636 | Missense Mutation | AAG,AGG | K,R 113 | XP_011530198.1 |