Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCAGAGCATTTCGGTATATTGCAT[A/G]TATCTTCTGTTAAAAGAAAAACTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SCOC PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SCOC - short coiled-coil protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001153446.1 | Intron | NP_001146918.1 | ||||
NM_001153484.1 | Intron | NP_001146956.1 | ||||
NM_001153552.1 | Intron | NP_001147024.1 | ||||
NM_001153585.1 | Intron | NP_001147057.1 | ||||
NM_001153635.1 | Intron | NP_001147107.1 | ||||
NM_001153663.1 | Intron | NP_001147135.1 | ||||
NM_001153690.1 | Intron | NP_001147162.1 | ||||
NM_032547.2 | Intron | NP_115936.2 |