Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTCTGTAAACTGCTCCATCAGCA[C/G]GGTCCATTTCGAATGATTTTAATAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605753 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPINK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPINK2 - serine peptidase inhibitor, Kazal type 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271718.1 | 570 | Silent Mutation | CCC,CCG | P,P 133 | NP_001258647.1 | |
NM_001271719.1 | 570 | UTR 3 | NP_001258648.1 | |||
NM_001271720.1 | 570 | Silent Mutation | CCC,CCG | P,P 118 | NP_001258649.1 | |
NM_001271721.1 | 570 | UTR 3 | NP_001258650.1 | |||
NM_001271722.1 | 570 | Missense Mutation | CCT,CGT | P,R 82 | NP_001258651.1 | |
NM_021114.3 | 570 | Silent Mutation | CCC,CCG | P,P 83 | NP_066937.1 | |
XM_011534405.1 | 570 | Silent Mutation | CCC,CCG | P,P 185 | XP_011532707.1 | |
XM_011534406.2 | 570 | Intron | XP_011532708.1 |