Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTCTAACTGATGGTTGATATTTC[A/G]CAGATGGCTTTCGGCTCCCAGAAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611511 MIM: 615421 | ||||||||||||||||||||
Literature Links: |
CENPU PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CENPU - centromere protein U | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024629.3 | 1283 | Nonsense Mutation | CGA,TGA | R,* 405 | NP_078905.2 | |
XM_005263218.4 | 1283 | Nonsense Mutation | CGA,TGA | R,* 435 | XP_005263275.2 |