Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGTCCAATTGAATGGTTTCACTT[C/T]TCATGTGTTTCACTTACCTATAAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604215 | ||||||||||||||||||||
Literature Links: |
ING2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ING2 - inhibitor of growth family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291959.1 | 711 | Silent Mutation | TTC,TTT | F,F 200 | NP_001278888.1 | |
NM_001564.3 | 711 | Silent Mutation | TTC,TTT | F,F 240 | NP_001555.1 | |
XM_011531927.2 | 711 | Silent Mutation | TTC,TTT | F,F 185 | XP_011530229.1 |
LOC389247 - uncharacterized LOC389247 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |