Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAGCAGCTCACCACAGGGAAAAA[C/G]AGCATCTGCAAAAACGATGTCAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603064 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105377267 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105377267 - uncharacterized LOC105377267 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UGT2B11 - UDP glucuronosyltransferase family 2 member B11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011531550.2 | Intron | XP_011529852.1 | ||||
XM_017007660.1 | Intron | XP_016863149.1 | ||||
XM_017007661.1 | Intron | XP_016863150.1 |