Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCAACCCTTACCTTTGTTTTGCA[C/T]GGAGTATTCTCAGAGGTCACACTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614682 | ||||||||||||||||||||
Literature Links: |
ETNPPL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ETNPPL - ethanolamine-phosphate phospho-lyase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146590.1 | 1443 | Silent Mutation | CCA,CCG | P,P 447 | NP_001140062.1 | |
NM_001146627.1 | 1443 | Silent Mutation | CCA,CCG | P,P 395 | NP_001140099.1 | |
NM_031279.3 | 1443 | Silent Mutation | CCA,CCG | P,P 453 | NP_112569.2 | |
XM_006714290.2 | 1443 | Silent Mutation | CCA,CCG | P,P 413 | XP_006714353.1 |