Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAGTAGCTGGAGCCGGAGCAGGG[A/T]CAGAACCTGTTGCTGCAGACGGGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607449 | ||||||||||||||||||||
Literature Links: |
NPFFR2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NPFFR2 - neuropeptide FF receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144756.1 | 328 | Intron | NP_001138228.1 | |||
NM_004885.2 | 328 | Missense Mutation | GAC,GTC | D,V 77 | NP_004876.2 | |
NM_053036.2 | 328 | Intron | NP_444264.1 | |||
XM_011531554.2 | 328 | Intron | XP_011529856.1 |