Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAAGCAGCGGTCCCGGCCGCTGTC[C/T]CGCAGGCAGTAAAAGCCCTTGGTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603244 | ||||||||||||||||||||
Literature Links: |
HS3ST1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HS3ST1 - heparan sulfate-glucosamine 3-sulfotransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005114.2 | 5856 | Silent Mutation | CGA,CGG | R,R 258 | NP_005105.1 | |
XM_011513913.2 | 5856 | Silent Mutation | CGA,CGG | R,R 258 | XP_011512215.1 |