Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTCAAACTGCGAACGCCGCTTCC[C/T]ACTCCGTGCCATCGAAGAGAAGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606180 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EXOSC9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EXOSC9 - exosome component 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001034194.1 | 149 | Missense Mutation | CCA,CTA | P,L 14 | NP_001029366.1 | |
NM_005033.2 | 149 | Missense Mutation | CCA,CTA | P,L 14 | NP_005024.2 | |
XM_011532035.2 | 149 | Missense Mutation | CCA,CTA | P,L 14 | XP_011530337.1 |