Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATGTGGATGATTTCACCACTTTT[C/T]GTGGCACAGAGCTCAGCTTCCTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 131243 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EDNRA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EDNRA - endothelin receptor type A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166055.1 | 651 | Missense Mutation | CGT,TGT | R,C 41 | NP_001159527.1 | |
NM_001256283.1 | 651 | Intron | NP_001243212.1 | |||
NM_001957.3 | 651 | Missense Mutation | CGT,TGT | R,C 41 | NP_001948.1 |