Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATCGGGATGTGGTCAAGATGATC[A/G]CGGACACCAGCGAGGTCAAGTTGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606453 MIM: 604357 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC729558 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC729558 - uncharacterized LOC729558 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRBA - LPS responsive beige-like anchor protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199282.2 | 1562 | Intron | NP_001186211.2 | |||
NM_006726.4 | 1562 | Intron | NP_006717.2 | |||
XM_005263372.3 | 1562 | Intron | XP_005263429.1 | |||
XM_005263373.2 | 1562 | Intron | XP_005263430.1 | |||
XM_005263374.3 | 1562 | Intron | XP_005263431.1 | |||
XM_005263375.3 | 1562 | Intron | XP_005263432.1 | |||
XM_011532434.2 | 1562 | Intron | XP_011530736.1 | |||
XM_017008872.1 | 1562 | Intron | XP_016864361.1 | |||
XM_017008873.1 | 1562 | Intron | XP_016864362.1 | |||
XM_017008874.1 | 1562 | Intron | XP_016864363.1 |
MAB21L2 - mab-21 like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006439.4 | 1562 | Missense Mutation | ACG,GCG | T,A 153 | NP_006430.1 |