Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTAACTATTGGTACTTCAGAGATG[A/G]ATGCATTCTTGGATGACCCAGAATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612101 | ||||||||||||||||||||
Literature Links: |
PI4K2B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PI4K2B - phosphatidylinositol 4-kinase type 2 beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018323.3 | 423 | Missense Mutation | AAT,GAT | N,D 98 | NP_060793.2 | |
XM_005248174.1 | 423 | Missense Mutation | AAT,GAT | N,D 98 | XP_005248231.1 | |
XM_005248175.4 | 423 | Missense Mutation | AAT,GAT | N,D 2 | XP_005248232.1 |