Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATTTTCCAGTTTTCATTAATTCA[A/G]CTGCTTCATTGATTTTATCAAGATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 103735 | ||||||||||||||||||||
Literature Links: |
ADH6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADH6 - alcohol dehydrogenase 6 (class V) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000672.3 | 1173 | Missense Mutation | GCT,GTT | A,V 360 | NP_000663.1 | |
NM_001102470.1 | 1173 | Missense Mutation | GCT,GTT | A,V 360 | NP_001095940.1 |
LOC100507053 - uncharacterized LOC100507053 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |