Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGCGGCTGTAATTAAACACTTCA[A/G]GTGGACACCACTTCACAGGAAACTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600583 MIM: 600058 | ||||||||||||||||||||
Literature Links: |
TEC PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TEC - tec protein tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003215.2 | 2898 | Missense Mutation | CCT,CTT | P,L 534 | NP_003206.2 | |
XM_011513737.2 | 2898 | Missense Mutation | CCT,CTT | P,L 498 | XP_011512039.1 | |
XM_011513741.1 | 2898 | UTR 3 | XP_011512043.1 | |||
XM_017008569.1 | 2898 | Missense Mutation | CCT,CTT | P,L 418 | XP_016864058.1 |
TXK - TXK tyrosine kinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |