Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACAGCCGTCACTTACCCATGACC[A/G]TCACCTTCCTAAAGCTGTCTCGATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616047 MIM: 103220 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFAP97 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CFAP97 - cilia and flagella associated protein 97 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001292033.1 | 4525 | Intron | NP_001278962.1 | |||
NM_020827.2 | 4525 | UTR 3 | NP_065878.1 | |||
XM_017008483.1 | 4525 | UTR 3 | XP_016863972.1 | |||
XM_017008484.1 | 4525 | UTR 3 | XP_016863973.1 | |||
XM_017008485.1 | 4525 | UTR 3 | XP_016863974.1 | |||
XM_017008486.1 | 4525 | UTR 3 | XP_016863975.1 |
SLC25A4 - solute carrier family 25 member 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |