Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTATAAAGAACCTCTTAAGAAATG[C/T]ATCTTGTGTGGAAAGCATGTAGATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611143 MIM: 606769 MIM: 611983 | ||||||||||||||||||||
Literature Links: |
FAM175A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM175A - family with sequence similarity 175 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HELQ - helicase, POLQ-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPS18C - mitochondrial ribosomal protein S18C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297767.1 | 341 | Intron | NP_001284696.1 | |||
NM_001297769.1 | 341 | Silent Mutation | TGC,TGT | C,C 65 | NP_001284698.1 | |
NM_001297770.1 | 341 | Intron | NP_001284699.1 | |||
NM_016067.3 | 341 | Silent Mutation | TGC,TGT | C,C 65 | NP_057151.1 |