Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTGCCAGAAAGTCAAATGAACAA[A/G]CAACTTCTCTAAACACAGTTGGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601456 | ||||||||||||||||||||
Literature Links: |
MMRN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MMRN1 - multimerin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007351.2 | 553 | Missense Mutation | ACA,GCA | T,A 153 | NP_031377.2 | |
XM_005262856.2 | 553 | Missense Mutation | ACA,GCA | T,A 153 | XP_005262913.1 | |
XM_017007893.1 | 553 | Missense Mutation | ACA,GCA | T,A 153 | XP_016863382.1 |