Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCGTTCCTCCAGCAGAACTTCTAG[C/G]GGAGTTTGCCAACTACTTCCACTAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
HELT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HELT - helt bHLH transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300781.1 | 330 | Missense Mutation | GCG,GGG | A,G 81 | NP_001287710.1 | |
NM_001300782.1 | 330 | Missense Mutation | GCG,GGG | A,G 80 | NP_001287711.1 | |
XM_017008186.1 | 330 | Missense Mutation | GCG,GGG | A,G 43 | XP_016863675.1 |