Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAACACCTTCTGGATCTGGACTT[C/T]AGTCATGACCAGCCCCTCATAGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606173 MIM: 608790 | ||||||||||||||||||||
Literature Links: |
GRPEL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GRPEL1 - GrpE like 1, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025196.3 | 620 | Missense Mutation | AAA,GAA | K,E 148 | NP_079472.1 |
TADA2B - transcriptional adaptor 2B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |