Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCACACCCAACTATGAAGATCAG[C/T]ATCCTGCTCATGTTCCTTTGGGGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600980 | ||||||||||||||||||||
Literature Links: |
DMP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DMP1 - dentin matrix acidic phosphoprotein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079911.2 | 991 | Silent Mutation | AGC,AGT | S,S 4 | NP_001073380.1 | |
NM_004407.3 | 991 | Silent Mutation | AGC,AGT | S,S 4 | NP_004398.1 | |
XM_011531705.2 | 991 | Silent Mutation | AGC,AGT | S,S 33 | XP_011530007.1 | |
XM_011531706.2 | 991 | Silent Mutation | AGC,AGT | S,S 33 | XP_011530008.1 |