Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTCTAGGGCTTCTTGGTAATTCT[G/T]TTGCTATGCCAGTGAGTATTTTTTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606585 | ||||||||||||||||||||
Literature Links: |
ENAM PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ENAM - enamelin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031889.2 | 2142 | Missense Mutation | GTT,TTT | V,F 38 | NP_114095.2 | |
XM_006714056.3 | 2142 | Missense Mutation | GTT,TTT | V,F 38 | XP_006714119.1 |